Learn More
Ethos Biosciences Gram Stain, Safranin, 1gal, Ethos
Catalog No. 5000574258
Encompass_Preferred
Safranin is a staining solution suitable for microbiology, cytology, histology, and general laboratory applications. Commonly used as a counterstain in Gram staining procedures, it provides clear coloration of gram-negative organisms for reliable microscopic evaluation. The formulation delivers consistent color intensity, supporting accurate differentiation of cellular components. It is also suitable for general staining and research applications. Supplied in chemically compatible containers, Safranin is appropriate for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74258 Supplier Ethos Biosciences Supplier No. 6933G
Safranin is a staining solution suitable for microbiology, cytology, histology, and general laboratory applications. Commonly used as a counterstain in Gram staining procedures, it provides clear coloration of gram-negative organisms for reliable microscopic evaluation. The formulation delivers consistent color intensity, supporting accurate differentiation of cellular components. It is also suitable for general staining and research applications. Supplied in chemically compatible containers, Safranin is appropriate for clinical, research, and educational laboratory environments.