Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology SignalStain™ DAB Substrate Kit, Immunohistochemistry, 120 Slides, 1 Kit
SDP

Catalog No. NC1076811
Encompass

The SignalStain™ DAB Substrate Kit provides the reagents necessary to prepare a working solution of diaminobenzidine (DAB) for staining tissue sections. The working solution reacts with peroxidase (HRP) detection systems to yield a brown reaction product. This kit is suitable for use in immunohistochemistry (paraffin) applications.

  • Provides a brown reaction product with HRP-based detection systems.
  • Contains enough reagents to stain 120 slides.
  • Working solution is stable for up to 14 days at 4°C or up to 5 days at room temperature (approximately 25°C).
  • Shelf life of 12 months upon receipt when stored at 4°C and protected from light.
  • Requires standard laboratory safety precautions as DAB is a suspected carcinogen.
  • Includes DAB diluent and DAB chromogen concentrate components.

Catalog No. NC1076811 Supplier Cell Signaling Technology Supplier No. 8059P
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The SignalStain™ DAB Substrate Kit provides the reagents necessary to prepare a working solution of diaminobenzidine (DAB) for staining tissue sections. The working solution reacts with peroxidase (HRP) detection systems to yield a brown reaction product. This kit is suitable for use in immunohistochemistry (paraffin) applications.

  • Provides a brown reaction product with HRP-based detection systems.
  • Contains enough reagents to stain 120 slides.
  • Working solution is stable for up to 14 days at 4°C or up to 5 days at room temperature (approximately 25°C).
  • Shelf life of 12 months upon receipt when stored at 4°C and protected from light.
  • Requires standard laboratory safety precautions as DAB is a suspected carcinogen.
  • Includes DAB diluent and DAB chromogen concentrate components.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.