Learn More
Med Vet International Dip Quick Stain Kit Refill 3X500mL
Catalog No. 502831949
Encompass_Preferred
Excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds.500ml Refill Kit. 500ml of fixative, eosin and counter stain.
Catalog No. 50-283-1949 Supplier Med Vet International Supplier No. JOR322A
Excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds.500ml Refill Kit. 500ml of fixative, eosin and counter stain.