Learn More
Med Vet International Jorvet Dip Quick Stain #2, Eosin Only, 500mL, J0322A2
Catalog No. 502829420
Encompass_Preferred
The Dip Quick Stain #2 Eosin Solution is recommended use for invitro veterinary use only. It is excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds.
Catalog No. 50-282-9420 Supplier Med Vet International Supplier No. JOR322A2
The Dip Quick Stain #2 Eosin Solution is recommended use for invitro veterinary use only. It is excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds.