Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Stain Dish with Cover
SDP

Catalog No. 21010249
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-010-249 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-010-249 Supplier StatLab Supplier No. 900303EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stain Dish with Cover is made from soda lime glass.

  • Accommodates 25-30 slide rack
  • 118 x 88 x 70 mm

Specifications

Autoclavable Autoclavable
Bottom Shape Flat
Capacity (Metric) 0.727 L
Color Clear
Material Glass
Type Staining Dish
Dimensions (L x W x H) 118 x 88 x 70 mm
Height (Metric) 70 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.