Learn More
Med Vet International Stain Dish, Rectangular
Catalog No. 503045629
Encompass_Preferred
Glass cover, holds 20 slides (back to back). Inside dimensions: 3" x 2 1/4in. x 1 1/4in.. Individually boxed,
Catalog No. 50-304-5629 Supplier Med Vet International Supplier No. SD10
Glass cover, holds 20 slides (back to back). Inside dimensions: 3" x 2 1/4in. x 1 1/4in.. Individually boxed,