Learn More
Med Vet International Stain Dish, Coplin Jar
Catalog No. 502832019
Encompass_Preferred
Ground-on glass cover and wide, flat, round base. 3" high, inside diameter 1.2" Capacity: 10 slides (back to back). Individually boxed
Catalog No. 50-283-2019 Supplier Med Vet International Supplier No. SD05
Ground-on glass cover and wide, flat, round base. 3" high, inside diameter 1.2" Capacity: 10 slides (back to back). Individually boxed