Learn More
Newcomer Supply Iron, Gomori Prussian Blue Stain Kit, 250 ml kit
SDP
Catalog No. NC0777941
Encompass
Newcomer Supply Iron, Gomori Prussian Blue Stain Kit procedure is used to detect loosely bound ferric iron in tissue sections. This histochemical reaction is sensitive enough to demonstrate minute amounts of iron deposits in blood cells, bone marrow and spleen.
Catalog No. NC0777941 Supplier Newcomer Supply Supplier No. 9136A
Newcomer Supply Iron, Gomori Prussian Blue Stain Kit procedure is used to detect loosely bound ferric iron in tissue sections. This histochemical reaction is sensitive enough to demonstrate minute amounts of iron deposits in blood cells, bone marrow and spleen.