Learn More
Ethos Biosciences Wright's Stain, 16oz, Ethos
Catalog No. 5000574182
Encompass_Preferred
Wright Stain is a Romanowsky-type staining solution designed for hematology, cytology, and microbiology applications. Commonly used for staining peripheral blood smears and bone marrow specimens, it provides clear differentiation of cellular components for reliable microscopic evaluation. The formulation delivers consistent nuclear and cytoplasmic staining, supporting accurate cell morphology assessment and differential counts. Ideal for routine diagnostic workflows and research procedures, Wright Stain offers dependable and reproducible results.
Catalog No. 50-005-74182 Supplier Ethos Biosciences Supplier No. 558016
Wright Stain is a Romanowsky-type staining solution designed for hematology, cytology, and microbiology applications. Commonly used for staining peripheral blood smears and bone marrow specimens, it provides clear differentiation of cellular components for reliable microscopic evaluation. The formulation delivers consistent nuclear and cytoplasmic staining, supporting accurate cell morphology assessment and differential counts. Ideal for routine diagnostic workflows and research procedures, Wright Stain offers dependable and reproducible results.