Learn More
Ethos Biosciences Quick I Red, 32oz, Ethos
Catalog No. 5000574236
Encompass_Preferred
ready-to-use, two-step, staining solution replicating Wright Stain designed for hematology and cytology. Commonly used for rapid staining of peripheral blood smears and bone marrow specimens, it provides clear differentiation of cellular components for reliable microscopic evaluation. The formulation delivers consistent color intensity and rapid results, making it ideal for routine diagnostic workflows and high-throughput laboratories. Suitable for clinical, research, and educational settings, Quick I Red supports accurate cell morphology assessment and dependable staining performance.
Catalog No. 50-005-74236 Supplier Ethos Biosciences Supplier No. 5131R32
ready-to-use, two-step, staining solution replicating Wright Stain designed for hematology and cytology. Commonly used for rapid staining of peripheral blood smears and bone marrow specimens, it provides clear differentiation of cellular components for reliable microscopic evaluation. The formulation delivers consistent color intensity and rapid results, making it ideal for routine diagnostic workflows and high-throughput laboratories. Suitable for clinical, research, and educational settings, Quick I Red supports accurate cell morphology assessment and dependable staining performance.