Learn More
Ethos Biosciences Toluidine Blue 1%, 16oz, Ethos
Catalog No. 5000574151
Encompass_Preferred
Toluidine Blue 1% is a ready-to-use staining solution suitable for histology, cytology, microbiology, and general laboratory applications. Commonly used as a basic dye, it provides strong nuclear and metachromatic staining, allowing clear visualization of cellular and tissue components during microscopic evaluation. The formulation delivers consistent color intensity for reliable slide preparation and reproducible results. Ideal for routine and specialized staining protocols, Toluidine Blue 1% supports accurate cell and tissue morphology assessment.
Catalog No. 50-005-74151 Supplier Ethos Biosciences Supplier No. 605416
Toluidine Blue 1% is a ready-to-use staining solution suitable for histology, cytology, microbiology, and general laboratory applications. Commonly used as a basic dye, it provides strong nuclear and metachromatic staining, allowing clear visualization of cellular and tissue components during microscopic evaluation. The formulation delivers consistent color intensity for reliable slide preparation and reproducible results. Ideal for routine and specialized staining protocols, Toluidine Blue 1% supports accurate cell and tissue morphology assessment.