Learn More
Ethos Biosciences Alcian Blue 1%, pH 2.5 Stain Kit, 3x16oz, Ethos
Catalog No. 5000574119
Encompass_Preferred
Alcian Blue pH 2.5 Stain is a ready-to-use solution formulated for the demonstration of acidic mucopolysaccharides and glycosaminoglycans in tissue sections. At pH 2.5, the stain selectively colors both sulfated and carboxylated acidic mucins, providing clear blue staining for reliable visualization and contrast with nuclear counterstains. Commonly used in histology, pathology, and research laboratories, it supports routine special staining procedures and teaching applications. Compatible with standard Alcian Blue staining protocols, this solution delivers consistent, reproducible results for the evaluation of mucin-containing tissues.
Catalog No. 50-005-74119 Supplier Ethos Biosciences Supplier No. 757616
Alcian Blue pH 2.5 Stain is a ready-to-use solution formulated for the demonstration of acidic mucopolysaccharides and glycosaminoglycans in tissue sections. At pH 2.5, the stain selectively colors both sulfated and carboxylated acidic mucins, providing clear blue staining for reliable visualization and contrast with nuclear counterstains. Commonly used in histology, pathology, and research laboratories, it supports routine special staining procedures and teaching applications. Compatible with standard Alcian Blue staining protocols, this solution delivers consistent, reproducible results for the evaluation of mucin-containing tissues.