Learn More
Ethos Biosciences Gill 2X Hematoxylin, 1gal, Ethos
Catalog No. 5000574135
Encompass_Preferred
Gill 2X Hematoxylin is a nuclear staining solution designed for histology and cytology applications. Formulated for progressive staining, it provides strong, crisp nuclear detail without the need for differentiation steps. The 2X formulation offers increased staining intensity compared to lower concentrations, making it suitable for routine diagnostic procedures and a wide range of tissue types. It delivers consistent color development for reliable microscopic evaluation and reproducible results. Suitable for clinical, research, and educational laboratories, Gill 2X Hematoxylin supports efficient and dependable staining workflows. Supplied in chemically compatible containers for convenient laboratory use.
Catalog No. 50-005-74135 Supplier Ethos Biosciences Supplier No. 7013G
Gill 2X Hematoxylin is a nuclear staining solution designed for histology and cytology applications. Formulated for progressive staining, it provides strong, crisp nuclear detail without the need for differentiation steps. The 2X formulation offers increased staining intensity compared to lower concentrations, making it suitable for routine diagnostic procedures and a wide range of tissue types. It delivers consistent color development for reliable microscopic evaluation and reproducible results. Suitable for clinical, research, and educational laboratories, Gill 2X Hematoxylin supports efficient and dependable staining workflows. Supplied in chemically compatible containers for convenient laboratory use.