Learn More
Ethos Biosciences Gill 1X Hematoxylin, 1gal, Ethos
Catalog No. 5000574111
Encompass_Preferred
Gill 1X Hematoxylin is a nuclear staining solution designed for histology and cytology applications. Formulated for progressive staining, it provides clear, crisp nuclear detail without the need for differentiation steps. The 1X formulation offers controlled staining intensity, making it ideal for delicate tissues and routine cytology diagnostic procedures. It delivers consistent color development for reliable microscopic evaluation and reproducible results. Suitable for clinical, research, and educational laboratories, Gill 1X Hematoxylin supports efficient and dependable staining workflows. Supplied in chemically compatible containers for convenient laboratory use.
Catalog No. 50-005-74111 Supplier Ethos Biosciences Supplier No. 7012G
Gill 1X Hematoxylin is a nuclear staining solution designed for histology and cytology applications. Formulated for progressive staining, it provides clear, crisp nuclear detail without the need for differentiation steps. The 1X formulation offers controlled staining intensity, making it ideal for delicate tissues and routine cytology diagnostic procedures. It delivers consistent color development for reliable microscopic evaluation and reproducible results. Suitable for clinical, research, and educational laboratories, Gill 1X Hematoxylin supports efficient and dependable staining workflows. Supplied in chemically compatible containers for convenient laboratory use.