Learn More
Ethos Biosciences Mayer's Hematoxylin, 16oz, Ethos
Catalog No. 5000574112
Encompass_Preferred
Mayer’s Hematoxylin is a nuclear staining solution designed for histology applications. Formulated as a progressive stain, it provides clear, crisp nuclear detail without the need for differentiation steps. The gentle staining action makes it ideal for delicate tissues and routine procedures, including H&E and special staining protocols. The formulation delivers consistent color development for reliable microscopic evaluation and reproducible results. Suitable for clinical, research, and educational laboratories, Mayer’s Hematoxylin supports efficient and dependable staining workflows. Supplied in chemically compatible containers for convenient laboratory use.
Catalog No. 50-005-74112 Supplier Ethos Biosciences Supplier No. 702116
Mayer’s Hematoxylin is a nuclear staining solution designed for histology applications. Formulated as a progressive stain, it provides clear, crisp nuclear detail without the need for differentiation steps. The gentle staining action makes it ideal for delicate tissues and routine procedures, including H&E and special staining protocols. The formulation delivers consistent color development for reliable microscopic evaluation and reproducible results. Suitable for clinical, research, and educational laboratories, Mayer’s Hematoxylin supports efficient and dependable staining workflows. Supplied in chemically compatible containers for convenient laboratory use.