Learn More
Med Vet International Miltex Surgical Instrument Stain Remover, 3oz, 12/Case
Catalog No. 502643541
Encompass_Preferred
The Miltex 3-740 3oz. Surgical Instrument Stainl Remover is a special powdered fromula that removes stains and tarnsihes to restore luster to surgical instruments. The Miltex Stain remover may also be used on autoclaves, ultrasonic cleaners, and stainless steel countertops.
Catalog No. 50-264-3541 Supplier Med Vet International Supplier No. MIL3740
The Miltex 3-740 3oz. Surgical Instrument Stainl Remover is a special powdered fromula that removes stains and tarnsihes to restore luster to surgical instruments. The Miltex Stain remover may also be used on autoclaves, ultrasonic cleaners, and stainless steel countertops.