Learn More
Ethos Biosciences AFB Kinyoun Methylene Blue, 4x8oz, Ethos
Catalog No. 5000574290
Encompass_Preferred
AFB Methylene Blue Counterstain is formulated for use as a counterstain in acid-fast staining procedures, including the Kinyoun and Ziehl-Neelsen methods. It provides a clear blue background that enhances contrast with the red-stained acid-fast organisms, supporting accurate microscopic differentiation. Designed for routine use in clinical, veterinary, and research laboratories, this counterstain is compatible with standard acid-fast staining protocols and manual procedures. The formulation delivers consistent color development and reliable performance for dependable staining results in microbiology applications.
Catalog No. 50-005-74290 Supplier Ethos Biosciences Supplier No. 6305
AFB Methylene Blue Counterstain is formulated for use as a counterstain in acid-fast staining procedures, including the Kinyoun and Ziehl-Neelsen methods. It provides a clear blue background that enhances contrast with the red-stained acid-fast organisms, supporting accurate microscopic differentiation. Designed for routine use in clinical, veterinary, and research laboratories, this counterstain is compatible with standard acid-fast staining protocols and manual procedures. The formulation delivers consistent color development and reliable performance for dependable staining results in microbiology applications.