Learn More
Ethos Biosciences AFB Kinyoun Stain Kit Green, 3x8oz, Ethos
Catalog No. 5000574159
Encompass_Preferred
AFB Kinyoun Stain Kit with Brilliant Green Counterstain is a complete cold staining system designed for the detection of acid-fast organisms in clinical and research laboratories. Based on the Kinyoun method, this kit uses a concentrated carbol fuchsin primary stain, acid-alcohol decolorizer, and brilliant green counterstain to provide strong contrast between acid-fast bacteria and background material. Acid-fast organisms appear bright red against a green background, supporting clear visualization and differentiation. Suitable for manual staining procedures, the kit is compatible with standard Kinyoun protocols and provides consistent, reproducible results for routine microbiology applications.
Catalog No. 50-005-74159 Supplier Ethos Biosciences Supplier No. 630708
AFB Kinyoun Stain Kit with Brilliant Green Counterstain is a complete cold staining system designed for the detection of acid-fast organisms in clinical and research laboratories. Based on the Kinyoun method, this kit uses a concentrated carbol fuchsin primary stain, acid-alcohol decolorizer, and brilliant green counterstain to provide strong contrast between acid-fast bacteria and background material. Acid-fast organisms appear bright red against a green background, supporting clear visualization and differentiation. Suitable for manual staining procedures, the kit is compatible with standard Kinyoun protocols and provides consistent, reproducible results for routine microbiology applications.