Learn More
Ethos Biosciences AFB Ziehl Neelsen Methylene Blue, 8oz, Ethos
Catalog No. 5000574162
Encompass_Preferred
AFB Methylene Blue Counterstain is formulated for use in acid-fast staining procedures, including the Ziehl–Neelsen and Kinyoun methods. Applied after decolorization, it provides a clean blue background that enhances contrast with red-stained acid-fast organisms, supporting clear microscopic differentiation. Designed for routine use in clinical, veterinary, and research laboratories, this counterstain is compatible with standard acid-fast staining protocols and manual procedures. The formulation delivers consistent color development and dependable performance for reliable microbiology staining results.
Catalog No. 50-005-74162 Supplier Ethos Biosciences Supplier No. 633008
AFB Methylene Blue Counterstain is formulated for use in acid-fast staining procedures, including the Ziehl–Neelsen and Kinyoun methods. Applied after decolorization, it provides a clean blue background that enhances contrast with red-stained acid-fast organisms, supporting clear microscopic differentiation. Designed for routine use in clinical, veterinary, and research laboratories, this counterstain is compatible with standard acid-fast staining protocols and manual procedures. The formulation delivers consistent color development and dependable performance for reliable microbiology staining results.