Learn More
Ethos Biosciences AFB Brilliant Green, Kinyoun, 4x8oz, Ethos
Catalog No. 5000574208
Encompass_Preferred
AFB Brilliant Green Counterstain is designed for use in acid-fast staining procedures, including the Kinyoun method, to provide strong contrast between acid-fast organisms and background material. When used after carbol fuchsin staining and decolorization, the counterstain produces a clean green background, allowing acid-fast bacteria to appear bright red for clear visualization. Suitable for routine use in clinical, veterinary, and research laboratories, this reagent is compatible with standard acid-fast staining protocols and manual procedures. The formulation supports consistent color development and reliable results in microbiology applications.
Catalog No. 50-005-74208 Supplier Ethos Biosciences Supplier No. 6303
AFB Brilliant Green Counterstain is designed for use in acid-fast staining procedures, including the Kinyoun method, to provide strong contrast between acid-fast organisms and background material. When used after carbol fuchsin staining and decolorization, the counterstain produces a clean green background, allowing acid-fast bacteria to appear bright red for clear visualization. Suitable for routine use in clinical, veterinary, and research laboratories, this reagent is compatible with standard acid-fast staining protocols and manual procedures. The formulation supports consistent color development and reliable results in microbiology applications.