Learn More
Ethos Biosciences AFB Kinyoun Stain Kit Blue, 3x8oz, Ethos
Catalog No. 5000574357
Encompass_Preferred
AFB Kinyoun Stain Kit with Methylene Blue Counterstain is a complete cold staining system designed for the detection of acid-fast organisms in clinical and research laboratories. Based on the Kinyoun method, this kit includes a concentrated carbol fuchsin primary stain, acid-alcohol decolorizer, and methylene blue counterstain to provide clear contrast between acid-fast bacteria and background material. Acid-fast organisms appear bright red against a blue background, supporting reliable visualization and differentiation. Suitable for manual staining procedures, the kit is compatible with standard Kinyoun protocols and delivers consistent, reproducible results for routine microbiology applications.
Catalog No. 50-005-74357 Supplier Ethos Biosciences Supplier No. 630608
AFB Kinyoun Stain Kit with Methylene Blue Counterstain is a complete cold staining system designed for the detection of acid-fast organisms in clinical and research laboratories. Based on the Kinyoun method, this kit includes a concentrated carbol fuchsin primary stain, acid-alcohol decolorizer, and methylene blue counterstain to provide clear contrast between acid-fast bacteria and background material. Acid-fast organisms appear bright red against a blue background, supporting reliable visualization and differentiation. Suitable for manual staining procedures, the kit is compatible with standard Kinyoun protocols and delivers consistent, reproducible results for routine microbiology applications.