Learn More
Ethos Biosciences Malachite Green 1%, 16oz, Ethos
Catalog No. 5000574358
Encompass_Preferred
Malachite Green 1% is a ready-to-use staining solution formulated for microbiology and general laboratory applications. Commonly used as a counterstain in spore staining procedures, including the Schaeffer–Fulton method, it provides strong green coloration of background material while allowing spores stained with primary dyes to be clearly visualized. The formulation delivers consistent color intensity and reliable contrast for accurate microscopic evaluation. Suitable for use in clinical, research, and educational laboratories, Malachite Green 1% is compatible with standard staining protocols and manual procedures, supporting dependable and reproducible results.
Catalog No. 50-005-74358 Supplier Ethos Biosciences Supplier No. 633516
Malachite Green 1% is a ready-to-use staining solution formulated for microbiology and general laboratory applications. Commonly used as a counterstain in spore staining procedures, including the Schaeffer–Fulton method, it provides strong green coloration of background material while allowing spores stained with primary dyes to be clearly visualized. The formulation delivers consistent color intensity and reliable contrast for accurate microscopic evaluation. Suitable for use in clinical, research, and educational laboratories, Malachite Green 1% is compatible with standard staining protocols and manual procedures, supporting dependable and reproducible results.