Learn More
Ethos Biosciences Gentian Violet 1%, 16oz, Ethos
Catalog No. 5000574241
Encompass_Preferred
Gentian Violet 1% is a staining solution suitable for microbiology, cytology, and general laboratory applications. Commonly used as a primary stain in Gram staining procedures, it provides strong coloration of gram-positive organisms for reliable microscopic evaluation. The formulation delivers consistent color intensity, supporting accurate identification and differentiation of microorganisms. It is also suitable for general staining and research applications.
Catalog No. 50-005-74241 Supplier Ethos Biosciences Supplier No. 609016
Gentian Violet 1% is a staining solution suitable for microbiology, cytology, and general laboratory applications. Commonly used as a primary stain in Gram staining procedures, it provides strong coloration of gram-positive organisms for reliable microscopic evaluation. The formulation delivers consistent color intensity, supporting accurate identification and differentiation of microorganisms. It is also suitable for general staining and research applications.