Learn More
Ethos Biosciences AFB Kinyoun Carbol Fuchsin Replacement Kit, 4x8oz, Ethos
Catalog No. 5000574188
Encompass_Preferred
AFB Kinyoun Carbol Fuchsin is formulated for use as the primary stain in acid-fast staining procedures, including the Kinyoun “cold” method. It penetrates the waxy cell walls of acid-fast organisms, producing a strong red coloration that resists decolorization with acid-alcohol. This allows clear differentiation of acid-fast bacteria from non–acid-fast material after application of a suitable counterstain. Suitable for routine use in clinical, veterinary, and research laboratories, the reagent is compatible with standard acid-fast staining protocols and manual procedures. The formulation supports consistent staining performance and dependable results in microbiology applications.
Catalog No. 50-005-74188 Supplier Ethos Biosciences Supplier No. 6301
AFB Kinyoun Carbol Fuchsin is formulated for use as the primary stain in acid-fast staining procedures, including the Kinyoun “cold” method. It penetrates the waxy cell walls of acid-fast organisms, producing a strong red coloration that resists decolorization with acid-alcohol. This allows clear differentiation of acid-fast bacteria from non–acid-fast material after application of a suitable counterstain. Suitable for routine use in clinical, veterinary, and research laboratories, the reagent is compatible with standard acid-fast staining protocols and manual procedures. The formulation supports consistent staining performance and dependable results in microbiology applications.