Learn More
Ethos Biosciences Methylene Blue 1%, 4oz, Ethos
Catalog No. 5000574273
Encompass_Preferred
Methylene Blue 1% is a staining solution designed for cell staining and counterstaining in microbiology, cytology, and general laboratory applications. Commonly used as a simple stain or as a counterstain in various staining procedures, it provides clear contrast of cellular structures for reliable microscopic evaluation. The formulation delivers consistent color intensity, supporting accurate visualization of cell morphology and cellular detail. Ideal for routine staining protocols and research workflows, Methylene Blue 1% offers dependable and reproducible results. Supplied in chemically compatible containers, it is suitable for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74273 Supplier Ethos Biosciences Supplier No. 655004
Methylene Blue 1% is a staining solution designed for cell staining and counterstaining in microbiology, cytology, and general laboratory applications. Commonly used as a simple stain or as a counterstain in various staining procedures, it provides clear contrast of cellular structures for reliable microscopic evaluation. The formulation delivers consistent color intensity, supporting accurate visualization of cell morphology and cellular detail. Ideal for routine staining protocols and research workflows, Methylene Blue 1% offers dependable and reproducible results. Supplied in chemically compatible containers, it is suitable for clinical, research, and educational laboratory environments.