Learn More
Ethos Biosciences Loeffler's Methylene Blue, 4oz, Ethos
Catalog No. 5000574128
Encompass_Preferred
Loeffler’s Methylene Blue is a staining solution designed for microbiology, cytology, and general laboratory applications. Commonly used as a simple stain, it provides rapid coloration of bacterial cells for clear visualization of morphology during microscopic evaluation. The formulation delivers consistent color intensity, making it ideal for routine diagnostic and teaching laboratories. It is also suitable for use as a counterstain in select staining procedures. Supplied in chemically compatible containers, Loeffler’s Methylene Blue is appropriate for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74128 Supplier Ethos Biosciences Supplier No. 655804
Add to Cart
Add to Cart
Loeffler’s Methylene Blue is a staining solution designed for microbiology, cytology, and general laboratory applications. Commonly used as a simple stain, it provides rapid coloration of bacterial cells for clear visualization of morphology during microscopic evaluation. The formulation delivers consistent color intensity, making it ideal for routine diagnostic and teaching laboratories. It is also suitable for use as a counterstain in select staining procedures. Supplied in chemically compatible containers, Loeffler’s Methylene Blue is appropriate for clinical, research, and educational laboratory environments.