Learn More
Electron Microscopy Sciences Stain-'N'-Temp Slide
SDP
Catalog No. 5034720
Encompass_Preferred
This device is used as an aid when programming a microwave oven to achieve an ideal target temperature of 20-100ul staining volumes. The tool is designed to give quantitative temperature information about the droplet during staining of tissue sections on electron microscope grids.
This unit is a 4-well "PTFE" coated glass slide, each well 8 mm in diameter which will maintain the uniform shape of the droplet; 4 liquid crystal temperature strips ranging from 35 to 50 degree are affixed to each well to monitor the microwave oven temperature.
Developed in collaboration with Dr. Gary Login, Beth Israel Hospital, Boston, MA
Catalog No. 50-347-20 Supplier Electron Microscopy Sciences Supplier No. 9703001
This device is used as an aid when programming a microwave oven to achieve an ideal target temperature of 20-100ul staining volumes. The tool is designed to give quantitative temperature information about the droplet during staining of tissue sections on electron microscope grids.
This unit is a 4-well "PTFE" coated glass slide, each well 8 mm in diameter which will maintain the uniform shape of the droplet; 4 liquid crystal temperature strips ranging from 35 to 50 degree are affixed to each well to monitor the microwave oven temperature.
Developed in collaboration with Dr. Gary Login, Beth Israel Hospital, Boston, MA