Learn More
GA International Stain-Proof Xylene-Resistant Labels 1.375" x 1.375" / 34.9mm x 34.9mm Rectangle White From -80°C to +149°C (-112°F to +300°F)
Catalog No. 5000571991
Encompass_Preferred
Thermal-transfer printable labels designed for microscope slides. These highly glossy labels repel stains and do not acquire colors from histological stains, such as hematoxylin or eosin. The labels and printed information are xylene-resistant when used in combination with our XAR-class of solvent-resistant ribbons. The print will resist xylene, toluene, alcohol, MEK, and other solvents and chemicals without needing additional protection. Ideal for histology lab applications.
Catalog No. 50-005-71991 Supplier GA International Supplier No. XST535C33WH
Thermal-transfer printable labels designed for microscope slides. These highly glossy labels repel stains and do not acquire colors from histological stains, such as hematoxylin or eosin. The labels and printed information are xylene-resistant when used in combination with our XAR-class of solvent-resistant ribbons. The print will resist xylene, toluene, alcohol, MEK, and other solvents and chemicals without needing additional protection. Ideal for histology lab applications.