Learn More
GA International Stain-Proof Xylene-Resistant Labels 3.5" x 2" / 88.9mm x 50.8mm Rectangle White From -80°C to +149°C (-112°F to +300°F)
Catalog No. 5000570888
Encompass_Preferred
Thermal-transfer printable labels designed for microscope slides. These highly glossy labels repel stains and do not acquire colors from histological stains, such as hematoxylin or eosin. The labels and printed information are xylene-resistant when used in combination with our XAR-class of solvent-resistant ribbons. The print will resist xylene, toluene, alcohol, MEK, and other solvents and chemicals without needing additional protection. Ideal for histology lab applications.
Catalog No. 50-005-70888 Supplier GA International Supplier No. XST282C31WH
Thermal-transfer printable labels designed for microscope slides. These highly glossy labels repel stains and do not acquire colors from histological stains, such as hematoxylin or eosin. The labels and printed information are xylene-resistant when used in combination with our XAR-class of solvent-resistant ribbons. The print will resist xylene, toluene, alcohol, MEK, and other solvents and chemicals without needing additional protection. Ideal for histology lab applications.