Learn More
Ethos Biosciences Wright-Giemsa Stain Pack, 1set, Ethos
Catalog No. 5000574370
Encompass_Preferred
Wright-Giemsa Stain Stain-Pack is a convenient, ready-to-use solution designed for reliable hematology staining of peripheral blood smears and bone marrow specimens. Formulated for clear differentiation of cellular components, it delivers crisp nuclear detail and accurate cytoplasmic staining to support microscopic evaluation and differential counts. The Stain-Pack format streamlines workflow by providing pre-measured reagents designed to fit in platen stain analyzers. Wright-Giemsa Stain Stain-Pack helps ensure reproducible results, quality control compliance, and efficient turnaround in high-demand laboratory settings.
Catalog No. 50-005-74370 Supplier Ethos Biosciences Supplier No. 1001
Wright-Giemsa Stain Stain-Pack is a convenient, ready-to-use solution designed for reliable hematology staining of peripheral blood smears and bone marrow specimens. Formulated for clear differentiation of cellular components, it delivers crisp nuclear detail and accurate cytoplasmic staining to support microscopic evaluation and differential counts. The Stain-Pack format streamlines workflow by providing pre-measured reagents designed to fit in platen stain analyzers. Wright-Giemsa Stain Stain-Pack helps ensure reproducible results, quality control compliance, and efficient turnaround in high-demand laboratory settings.