Learn More
Electron Microscopy Sciences EMS Slide Staining Tray Removable Handle 30 slides
SDP
Catalog No. 501931273
Encompass_Preferred
Stainless steel slide staining dishes and trays are ideal for routine and special histology staining procedures. The staining trays have removable handles. Accepts 3" W (76 mm) x 1", 1½", or 2" H slides.
Capacity: 30 slides
Catalog No. 50-193-1273 Supplier Electron Microscopy Sciences Supplier No. 7222005
Stainless steel slide staining dishes and trays are ideal for routine and special histology staining procedures. The staining trays have removable handles. Accepts 3" W (76 mm) x 1", 1½", or 2" H slides.
Capacity: 30 slides