Learn More
Newcomer Supply Trichrome, Liver Control Slides, 10/set
SDP
Catalog No. NC0827135
Encompass
Newcomer Supply Trichrome, Liver Control Slides are for the positive histochemical staining of connective tissue and to differentially demonstrate collagen and muscle fibers. Trichrome, Liver Control Slides contain sections of positive staining liver.
Catalog No. NC0827135 Supplier Newcomer Supply Supplier No. 4690A
Newcomer Supply Trichrome, Liver Control Slides are for the positive histochemical staining of connective tissue and to differentially demonstrate collagen and muscle fibers. Trichrome, Liver Control Slides contain sections of positive staining liver.