Learn More
Newcomer Supply Elastic, Verhoeff Stain Kit, 250 ml kit
SDP
Catalog No. NC0795653
Encompass
Newcomer Supply Elastic, Verhoeff Stain Kit procedure, commonly referred to as Verhoeff-Van Gieson (VVG) technique, is used to demonstrate pathologic changes in elastic fibers as well as demonstration of normal elastic tissue such as arteries and veins.
Catalog No. NC0795653 Supplier Newcomer Supply Supplier No. 9116A
Newcomer Supply Elastic, Verhoeff Stain Kit procedure, commonly referred to as Verhoeff-Van Gieson (VVG) technique, is used to demonstrate pathologic changes in elastic fibers as well as demonstration of normal elastic tissue such as arteries and veins.