Learn More
Andwin Scientific STAINING DISH/GREEN 12/PK
SDP
Catalog No. NC9651739
Encompass_Preferred
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant and is sold in a pack of 12 green dishes.
Catalog No. NC9651739 Supplier Andwin Scientific Supplier No. 6254112
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant and is sold in a pack of 12 green dishes.