Learn More
Electron Microscopy Sciences 20-Slide Staining Dish and Lid only, 3¾" x 3" x 2¼" (H)
SDP
Catalog No. 50931043
Encompass_Preferred
These glass staining dishes are made of a heavy glass with snug fit lids. Lids are included. Dishes have a gasket seal. Accomodates 20 slides.
I.D.: 3¾" x 3" x 2¼" (H)
Dimensions: 4-1/4" x 3⅝" x 3" (H)
Catalog No. 50-931-043 Supplier Electron Microscopy Sciences Supplier No. 71420DL
These glass staining dishes are made of a heavy glass with snug fit lids. Lids are included. Dishes have a gasket seal. Accomodates 20 slides.
I.D.: 3¾" x 3" x 2¼" (H)
Dimensions: 4-1/4" x 3⅝" x 3" (H)