Learn More
Electron Microscopy Sciences EMS Slide Staining Dish w/Cover 30 slides, 114 (L) x 89 (W) mm
SDP
Catalog No. 501931271
Encompass_Preferred
Stainless steel slide staining dishes and trays are ideal for routine and special histology staining procedures. Comes complete with a cover.
Capacity: 30 slides
Dimensions: 114 (L) x 89 (W) mm
Height: 76 mm
Catalog No. 50-193-1271 Supplier Electron Microscopy Sciences Supplier No. 7222003
Stainless steel slide staining dishes and trays are ideal for routine and special histology staining procedures. Comes complete with a cover.
Capacity: 30 slides
Dimensions: 114 (L) x 89 (W) mm
Height: 76 mm