Learn More
Electron Microscopy Sciences EMS Slide Staining Dish w/Cover 60 slides, 202 (L) x 95 (W) mm
SDP
Catalog No. 501931272
Encompass_Preferred
*Tray not included. Stainless steel slide staining dishes are ideal for routine and special histology staining procedures. Comes complete with a cover.
Capacity: 60 slides
Dimensions: 202 (L) x 95 (W) mm
Height: 76 mm
Catalog No. 50-193-1272 Supplier Electron Microscopy Sciences Supplier No. 7222004
*Tray not included. Stainless steel slide staining dishes are ideal for routine and special histology staining procedures. Comes complete with a cover.
Capacity: 60 slides
Dimensions: 202 (L) x 95 (W) mm
Height: 76 mm