Learn More
Research Products International Corp Staining Dish with Rack and Lid, Glass, Holds 50 Slides
SDP
Catalog No. 501977528
Encompass
This slotted rack holds 50 microscope slides, size 3" x 1", (75 mm x 25 mm), 3" x 1 1/2" (75mm x 38 mm), and 3" x 2" (75 mm x 51 mm). The rack is made of non-tarnishable stainless steel this is resistant to staining solutions. The handle is permanently attached, but hinged to permit closure of the dish and easy insertion and removal of the microscope slides.  The dish is manufactured from soda-lime glass. 
Approximate inside dimensions: 185 mm L x 88 mm W x 68 mm D.
Catalog No. 50-197-7528 Supplier Research Products International Corp Supplier No. 144201
This slotted rack holds 50 microscope slides, size 3" x 1", (75 mm x 25 mm), 3" x 1 1/2" (75mm x 38 mm), and 3" x 2" (75 mm x 51 mm). The rack is made of non-tarnishable stainless steel this is resistant to staining solutions. The handle is permanently attached, but hinged to permit closure of the dish and easy insertion and removal of the microscope slides.  The dish is manufactured from soda-lime glass. 
Approximate inside dimensions: 185 mm L x 88 mm W x 68 mm D.