Learn More
Electron Microscopy Sciences Staining Jar with Tall Ground Glass Lid
SDP
Catalog No. 5029017
Encompass_Preferred
An all glass jar which holds 5 to 10 standard slides (3"x1"). They can be used as a chromatography jar with a choice of 2 ground glass lids. The lids are ground to the flange of the jar which deters evaporation. The jar with a taller lid (70316-04) accommodates larger slides: 4"x1" (100 mmx25 mm). An all glass jar which holds 5 to 10 standard slides (3"x1") or larger slides: 4"x1" (100mmx25mm).
Catalog No. 50-290-17 Supplier Electron Microscopy Sciences Supplier No. 7031604
An all glass jar which holds 5 to 10 standard slides (3"x1"). They can be used as a chromatography jar with a choice of 2 ground glass lids. The lids are ground to the flange of the jar which deters evaporation. The jar with a taller lid (70316-04) accommodates larger slides: 4"x1" (100 mmx25 mm). An all glass jar which holds 5 to 10 standard slides (3"x1") or larger slides: 4"x1" (100mmx25mm).