Learn More
Electron Microscopy Sciences Staining Jar with Ground Glass Lid
SDP
Catalog No. 50366812
Encompass_Preferred
An all glass jar which holds 5 to 10 standard slides (3"x1"). They can be used as a chromatography jar with a choice of 2 ground glass lids. The lids are ground to the flange of the jar which deters evaporation. The jar with a taller lid (70316-04) accommodates larger slides: 4"x1" (100 mmx25 mm). An all glass jar which holds 5 to 10 standard slides (3"x1").
Catalog No. 50-366-812 Supplier Electron Microscopy Sciences Supplier No. 7031602
An all glass jar which holds 5 to 10 standard slides (3"x1"). They can be used as a chromatography jar with a choice of 2 ground glass lids. The lids are ground to the flange of the jar which deters evaporation. The jar with a taller lid (70316-04) accommodates larger slides: 4"x1" (100 mmx25 mm). An all glass jar which holds 5 to 10 standard slides (3"x1").