Learn More
Research Products International Corp Staining Rack, 20 Slide Capacity
SDP
Catalog No. 50998390
Encompass
The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size: 3" x 1" (75 x 25 mm), 3" x 1-1/2" (75 x 38 mm) and 3" x 2" (75 x 51 mm) sizes. Manufactured from soda-lime glass.
Catalog No. 50-998-390 Supplier Research Products International Corp Supplier No. 144204
The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size: 3" x 1" (75 x 25 mm), 3" x 1-1/2" (75 x 38 mm) and 3" x 2" (75 x 51 mm) sizes. Manufactured from soda-lime glass.