Learn More
Newcomer Supply Staining Rack for 20 Slide Staining Dish - Stainless Steel
SDP
Catalog No. NC2102244
Encompass
The Stainless Steel Staining Rack accommodates 20 horizontal slides. Outside Dimensions: 3 3/8" x 2 1/2" x 2 1/8" (H). Racks are also available in 2 other sizes to accommodate 30 and 60 slides.
This item is not returnable. View return policy
Catalog No. NC2102244 Supplier Newcomer Supply Supplier No. 6817A
The Stainless Steel Staining Rack accommodates 20 horizontal slides. Outside Dimensions: 3 3/8" x 2 1/2" x 2 1/8" (H). Racks are also available in 2 other sizes to accommodate 30 and 60 slides.