Learn More
Electron Microscopy Sciences Staining Rack, Polypropylene
SDP
Catalog No. 5028711
Encompass_Preferred
Made from polypropylene, autoclavable, accommodates up to 20 slides: 3" x 1" (76 x 26 mm). Comes with a removable snap-on handle, measures: 2-3/4" x 3-3/8" x 13/16" (70 x 86 x 21 mm) without handle
Catalog No. 50-287-11 Supplier Electron Microscopy Sciences Supplier No. 7032110
Made from polypropylene, autoclavable, accommodates up to 20 slides: 3" x 1" (76 x 26 mm). Comes with a removable snap-on handle, measures: 2-3/4" x 3-3/8" x 13/16" (70 x 86 x 21 mm) without handle