Learn More
Chemglass Life Sciences Staining Solution, 1mL, Midori Green Nucleic Acid
SDP
Catalog No. 501532252
Encompass
Staining Solution, 1mL, Midori Green Nucleic Acid. Midori Green Nucleic Acid Staining Solution is aand safe alternative to the traditional ethidium bromide(EtBr) stain for detecting dsDNA, ssDNA and RNA in agarose gels. Midori Green is just as sensitive as EtBr and the staining protocol is also very similar. Midori Green binds less aggressively to the DNA molecules, so fewer mutations occur during synthesis. Compared to EtBr, which is known as a strong mutagen, Midori Green causes much fewer mutations in the Ames test. Most importantly, it is not considered hazardous waste, can be disposed of according to standard laboratories procedures, and is stable for years. Non-toxic. Non-mutagenic. Non-carcinogenic. Excitation: 270 nm and 290 nm (fluorescence) 490nm (visible). Emission: 530 nm (fluorescence). Compatible with UV Transilluminator and Blue LED Transilluminator. Sensitive. 1ml bottle (20,000x). Can be stored at room temperature, protected from light.
Catalog No. 50-153-2252 Supplier Chemglass Life Sciences Supplier No. CLS1909G01
Staining Solution, 1mL, Midori Green Nucleic Acid. Midori Green Nucleic Acid Staining Solution is aand safe alternative to the traditional ethidium bromide(EtBr) stain for detecting dsDNA, ssDNA and RNA in agarose gels. Midori Green is just as sensitive as EtBr and the staining protocol is also very similar. Midori Green binds less aggressively to the DNA molecules, so fewer mutations occur during synthesis. Compared to EtBr, which is known as a strong mutagen, Midori Green causes much fewer mutations in the Ames test. Most importantly, it is not considered hazardous waste, can be disposed of according to standard laboratories procedures, and is stable for years. Non-toxic. Non-mutagenic. Non-carcinogenic. Excitation: 270 nm and 290 nm (fluorescence) 490nm (visible). Emission: 530 nm (fluorescence). Compatible with UV Transilluminator and Blue LED Transilluminator. Sensitive. 1ml bottle (20,000x). Can be stored at room temperature, protected from light.