Learn More
Medicus Health Autoclavable Slide Staining Station Dish with Lid
SDP
Catalog No. NC2669074
Encompass
Autoclavable Slide Staining Station Dis with Lid is resistant to temperature between 0°C and 120°C so they can be used with immunohistochemistry procedures and are great for histological, cytological and hematological staining procedures as well with total immersion for slides. Dishes can be sterilized by autoclave and used in the microwave.
Catalog No. NC2669074 Supplier Medicus Health Supplier No. 7353M3
Autoclavable Slide Staining Station Dis with Lid is resistant to temperature between 0°C and 120°C so they can be used with immunohistochemistry procedures and are great for histological, cytological and hematological staining procedures as well with total immersion for slides. Dishes can be sterilized by autoclave and used in the microwave.