Learn More
Cole-Parmer Cole-Parmer Hosecock Clamp, Regular, Stainless Steel
Supplier:  Cole-Parmer 0812986
Encompass_Preferred
Hosecocks have a convex clamping surface and rounded edges that prevent accidental cuts and tears of tubing. Adjustment screw with an oversized head provides one-handed flow regulation.
Catalog No. NC1451843