Learn More
W L Walker Co STAINLESS SAMPLER BOTTLE
Catalog No. NC0986543
Encompass_Preferred
W.L. Walker Bottle Sampler Stainless Steel 32 oz. • Use to take a running sample. • Bottle and cord are not included. • Available with narrow mouth plastic, or glass bottle. • Call for options.
Catalog No. NC0986543 Supplier W L Walker Co Supplier No. 40822
W.L. Walker Bottle Sampler Stainless Steel 32 oz. • Use to take a running sample. • Bottle and cord are not included. • Available with narrow mouth plastic, or glass bottle. • Call for options.