Learn More
Integra Biosciences Corp Stainless SS cuvette MC 30 -
SDP
Catalog No. 5000882048
Encompass
Stainless steel cuvette MEDIACLAVE 30 - for medium sterilization incl stainless steel guide tube (length 311mm) for temperature probe
Catalog No. 50-008-82048 Supplier Integra Biosciences Corp Supplier No. 136060
Stainless steel cuvette MEDIACLAVE 30 - for medium sterilization incl stainless steel guide tube (length 311mm) for temperature probe