Learn More
TA Instruments Stainless Steel, 12 mm Plate, NST, HR X0
Supplier:  TA Instruments 511120.946
Encompass_Preferred
Waters TA Instruments HR X0 Stainless Steel Upper Plate (12 mm, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only.
Catalog No. 50-005-80784