Learn More
Sigma Aldrich Stainless Steel - 17-7PH alloy, FeCr17Ni7
Catalog No. 503100276 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-310-0276 1 Ea.
1 options
Catalog No. 50-310-0276 Supplier Sigma Aldrich Supplier No. GF595623331EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Stainless Steel - 17-7PH alloy, FeCr17Ni7
Specifications
Quantity 1 Ea.
Synonym FE280321